Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114397.1 | NP_001107869.1 | 16 - 17 | 1959 | 105 |
NM_020865.2 | NP_065916.2 | 16 - 17 | 2001 | 105 | |
GenBank mRNA | AB040921.1 | - | 14 - 15 | 1453 | 105 |
AF217190.1 | - | 16 - 17 | 1993 | 105 | |
AJ577133.1 | - | 16 - 17 | 1920 | 105 | |
AJ577134.1 | - | 16 - 17 | 1878 | 105 | |
AK303720.1 | - | - | 1881 | 105 | |
AK314435.1 | - | 16 - 17 | 2001 | 105 | |
BC036035.1 | - | 16 - 17 | 1965 | 105 |