Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114397.1 | NP_001107869.1 | 6 - 7 | 976 | 118 |
NM_020865.2 | NP_065916.2 | 6 - 7 | 976 | 118 | |
GenBank mRNA | AB040921.1 | - | 4 - 5 | 428 | 118 |
AF217190.1 | - | 6 - 7 | 968 | 118 | |
AJ577133.1 | - | 6 - 7 | 895 | 118 | |
AJ577134.1 | - | 6 - 7 | 895 | 118 | |
AK303720.1 | - | - | 856 | 118 | |
AK314435.1 | - | 6 - 7 | 976 | 118 | |
BC036035.1 | - | 6 - 7 | 940 | 118 |