Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012452.2 | NP_036584.1 | 4 - 5 | 648 | 105 |
GenBank mRNA | AF023614.1 | - | 4 - 5 | 648 | 105 |
AK223453.1 | - | 4 - 5 | 648 | 105 | |
AK313302.1 | - | 4 - 5 | 668 | 105 | |
AX766384.1 | - | 4 - 5 | 648 | 105 | |
AX772740.1 | - | 3 - 4 | 502 | 105 | |
AY302137.1 | - | 3 - 4 | 497 | 105 | |
BC109392.1 | - | 3 - 4 | 509 | 105 |