Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277961.1 | NP_001264890.1 | 1 - 2 | 705 | 105 |
NM_001277962.1 | NP_001264891.1 | 1 - 2 | 705 | 105 | |
NM_003156.3 | NP_003147.2 | 1 - 2 | 705 | 105 | |
GenBank mRNA | AK314928.1 | - | 1 - 2 | 226 | 105 |
BC021300.2 | - | 1 - 2 | 697 | 105 | |
HQ317451.1 | - | 1 - 2 | 704 | 105 | |
JX014264.1 | - | 1 - 2 | 137 | 105 | |
U52426.1 | - | 1 - 2 | 702 | 105 |