Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277961.1 | NP_001264890.1 | 7 - 8 | 1537 | 75 |
NM_001277962.1 | NP_001264891.1 | 7 - 8 | 1537 | 75 | |
NM_003156.3 | NP_003147.2 | 7 - 8 | 1537 | 75 | |
GenBank mRNA | AK310510.1 | - | 4 - 5 | 663 | 75 |
AK314928.1 | - | 7 - 8 | 1058 | 75 | |
BC021300.2 | - | 7 - 8 | 1529 | 75 | |
HQ317451.1 | - | 7 - 8 | 1536 | 75 | |
JX014264.1 | - | 7 - 8 | 969 | 75 | |
U52426.1 | - | 7 - 8 | 1534 | 75 |