Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271838.1 | NP_001258767.1 | 3 - 4 | 493 | 105 |
NM_016625.3 | NP_057709.2 | 3 - 4 | 481 | 105 | |
GenBank mRNA | AF208853.1 | - | 3 - 4 | 439 | 105 |
AK290334.1 | - | 3 - 4 | 461 | 105 | |
BC006982.1 | - | 3 - 4 | 430 | 105 | |
DA273191.1 | - | 3 - 4 | 483 | 105 |