Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184743.1 | NP_001171672.1 | 2 - 3 | 836 | 73 |
NM_032507.3 | NP_115896.1 | 2 - 3 | 816 | 73 | |
XM_006715228.2 | XP_006715291.1 | 1 - 2 | 493 | 73 | |
XM_011514944.2 | XP_011513246.1 | 1 - 2 | 493 | 73 | |
XM_017011361.1 | XP_016866850.1 | 1 - 2 | 493 | 73 | |
XM_017011362.1 | XP_016866851.1 | 2 - 3 | 609 | 73 | |
XM_017011363.1 | XP_016866852.1 | 2 - 3 | 792 | 73 | |
GenBank mRNA | AK223446.1 | - | 2 - 3 | 801 | 73 |
AK291228.1 | - | 2 - 3 | 781 | 73 | |
BC032507.1 | - | 2 - 3 | 766 | 73 | |
BC069033.1 | - | 2 - 3 | 662 | 73 | |
BC128585.1 | - | 2 - 3 | 663 | 73 | |
D88259.1 | - | 1 - 2 | 396 | 73 |