Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021110.3 | NP_066933.1 | 13 - 14 | 1864 | 69 |
XM_005251059.3 | XP_005251116.1 | 13 - 14 | 1864 | 69 | |
XM_006716651.3 | XP_006716714.1 | 13 - 14 | 1727 | 69 | |
XM_017013809.1 | XP_016869298.1 | 13 - 14 | 1653 | 69 | |
GenBank mRNA | AK293080.1 | - | 13 - 14 | 1834 | 69 |
AK307716.1 | - | 13 - 14 | 1828 | 69 | |
AK311498.1 | - | 13 - 14 | 1828 | 69 | |
BC140893.1 | - | 12 - 13 | 1633 | 69 | |
BX472674.1 | - | 2 - 3 | 183 | 69 | |
M64108.1 | - | 8 - 9 | 1033 | 69 | |
M64109.1 | - | - | 865 | 69 |