Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145527.1 | NP_001138999.1 | 2 - 3 | 122 | 78 |
NM_024757.4 | NP_079033.4 | 2 - 3 | 122 | 78 | |
XM_005266105.4 | XP_005266162.1 | 2 - 3 | 165 | 78 | |
XM_006717288.2 | XP_006717351.1 | 2 - 3 | 83 | 78 | |
XM_011519021.2 | XP_011517323.1 | 2 - 3 | 1120 | 78 | |
XM_011519022.2 | XP_011517324.1 | 2 - 3 | 575 | 78 | |
XM_011519023.2 | XP_011517325.1 | 2 - 3 | 1120 | 78 | |
XM_011519024.2 | XP_011517326.1 | 2 - 3 | 1121 | 78 | |
XM_011519025.1 | XP_011517327.1 | 2 - 3 | 285 | 78 | |
XM_011519026.2 | XP_011517328.1 | 2 - 3 | 1122 | 78 | |
XM_011519027.2 | XP_011517329.1 | 2 - 3 | 1121 | 78 | |
XM_011519028.2 | XP_011517330.1 | 2 - 3 | 1121 | 78 | |
XM_011519033.2 | XP_011517335.1 | 2 - 3 | 1122 | 78 | |
XM_017015134.1 | XP_016870623.1 | 2 - 3 | 574 | 78 | |
XM_017015135.1 | XP_016870624.1 | 2 - 3 | 126 | 78 | |
XM_017015136.1 | XP_016870625.1 | 2 - 3 | 126 | 78 | |
XM_017015137.1 | XP_016870626.1 | 2 - 3 | 285 | 78 | |
GenBank mRNA | BC025772.1 | - | 2 - 3 | 112 | 78 |
BC047504.1 | - | 2 - 3 | 122 | 78 |