Assay Design
Gene Transcripts
- Gene Symbol
- STX1A
- Entrez Gene ID
-
6804
- Gene Name
- syntaxin 1A
- Gene Aliases
-
HPC-1, P35-1, STX1, SYN1A
- Chromosome Location
-
Chr.7: 73699205 - 73719702 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB086954.1 |
- |
8 - 9 |
676 |
76 |
View Full Product Details