Assay Design



Gene Transcripts

Gene Symbol
SUSD3
Entrez Gene ID
203328
Gene Name
sushi domain containing 3
Gene Aliases
-
Chromosome Location
Chr.9: 93058688 - 93085138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287005.1 NP_001273934.1 4 - 5 597 89
NM_001287008.1 NP_001273937.1 2 - 3 294 89
NM_145006.3 NP_659443.1 3 - 4 483 89
XM_011518358.1 XP_011516660.1 6 - 7 1677 89
XM_011518359.1 XP_011516661.1 5 - 6 1627 89
XM_011518361.2 XP_011516663.1 6 - 7 739 89
XM_011518362.1 XP_011516664.1 4 - 5 1513 89
XM_017014448.1 XP_016869937.1 4 - 5 784 89
XM_017014449.1 XP_016869938.1 3 - 4 665 89
XM_017014450.1 XP_016869939.1 5 - 6 624 89
XM_017014451.1 XP_016869940.1 3 - 4 1233 89
GenBank mRNA AI452970.1 - 2 - 3 203 89
AK128289.1 - 1 - 2 2451 89
AY358190.1 - 4 - 5 525 89
BC014601.2 - 3 - 4 464 89
BC041834.1 - 1 - 2 2377 89
DT220823.1 - 3 - 4 483 89

View Full Product Details