Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015150.1 | NP_055965.1 | 1 - 2 | 274 | 124 |
XM_011533529.1 | XP_011531831.1 | 1 - 2 | 226 | 124 | |
XM_011533531.1 | XP_011531833.1 | 1 - 2 | 226 | 124 | |
XM_017006005.1 | XP_016861494.1 | 1 - 2 | 226 | 124 | |
XM_017006006.1 | XP_016861495.1 | 1 - 2 | 226 | 124 | |
XM_017006007.1 | XP_016861496.1 | 1 - 2 | 258 | 124 | |
GenBank mRNA | AK074446.1 | - | 1 - 2 | 79 | 124 |
AU138240.1 | - | 1 - 2 | 335 | 124 | |
AY237654.1 | - | 1 - 2 | 265 | 124 | |
BC051336.1 | - | 1 - 2 | 51 | 124 | |
BC069209.1 | - | 1 - 2 | 51 | 124 | |
BC098270.1 | - | 1 - 2 | 98 | 124 | |
BC100667.1 | - | 1 - 2 | 98 | 124 | |
BC100669.1 | - | 1 - 2 | 98 | 124 | |
BC121795.1 | - | 1 - 2 | 98 | 124 | |
D42043.1 | - | 1 - 2 | 202 | 124 |