Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257971.1 | NP_001244900.1 | 5 - 6 | 881 | 87 |
NM_001257972.1 | NP_001244901.1 | 5 - 6 | 822 | 87 | |
NM_001257973.1 | NP_001244902.1 | 3 - 4 | 396 | 87 | |
NM_001912.4 | NP_001903.1 | 5 - 6 | 971 | 87 | |
NM_145918.2 | NP_666023.1 | 5 - 6 | 826 | 87 | |
XM_005251716.3 | XP_005251773.1 | 6 - 7 | 1701 | 87 | |
XM_011518263.1 | XP_011516565.1 | 5 - 6 | 1511 | 87 | |
XM_017014293.1 | XP_016869782.1 | 5 - 6 | 1448 | 87 | |
GenBank mRNA | AF217997.1 | - | 3 - 4 | 321 | 87 |
AK055599.1 | - | 5 - 6 | 1509 | 87 | |
AK075100.1 | - | 5 - 6 | 777 | 87 | |
AL832167.1 | - | 5 - 6 | 769 | 87 | |
BC012612.1 | - | 5 - 6 | 819 | 87 | |
BC142983.1 | - | 5 - 6 | 766 | 87 | |
BX537395.1 | - | 5 - 6 | 762 | 87 | |
BX647101.1 | - | 5 - 6 | 766 | 87 | |
BX647102.1 | - | 5 - 6 | 765 | 87 | |
BX647413.1 | - | 5 - 6 | 766 | 87 | |
BX647434.1 | - | 5 - 6 | 824 | 87 | |
BX647435.1 | - | 5 - 6 | 821 | 87 | |
BX648848.1 | - | 5 - 6 | 776 | 87 | |
BX648849.1 | - | 5 - 6 | 766 | 87 | |
BX649140.1 | - | 5 - 6 | 822 | 87 | |
CR457053.1 | - | 4 - 5 | 621 | 87 | |
X12451.1 | - | 5 - 6 | 909 | 87 | |
Y18462.1 | - | 2 - 3 | 246 | 87 |