Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257971.1 | NP_001244900.1 | 6 - 7 | 1046 | 79 |
NM_001257972.1 | NP_001244901.1 | 6 - 7 | 987 | 79 | |
NM_001257973.1 | NP_001244902.1 | 4 - 5 | 561 | 79 | |
NM_001912.4 | NP_001903.1 | 6 - 7 | 1136 | 79 | |
NM_145918.2 | NP_666023.1 | 6 - 7 | 991 | 79 | |
XM_005251716.3 | XP_005251773.1 | 7 - 8 | 1866 | 79 | |
GenBank mRNA | AF217997.1 | - | 4 - 5 | 486 | 79 |
AK055599.1 | - | 6 - 7 | 1674 | 79 | |
AK075100.1 | - | 6 - 7 | 942 | 79 | |
AL832167.1 | - | 6 - 7 | 934 | 79 | |
BC012612.1 | - | 6 - 7 | 984 | 79 | |
BC142983.1 | - | 6 - 7 | 931 | 79 | |
BM680778.1 | - | 2 - 3 | 79 | ||
BX537395.1 | - | 6 - 7 | 927 | 79 | |
BX647101.1 | - | 6 - 7 | 931 | 79 | |
BX647102.1 | - | 6 - 7 | 930 | 79 | |
BX647413.1 | - | 6 - 7 | 931 | 79 | |
BX647434.1 | - | 6 - 7 | 989 | 79 | |
BX647435.1 | - | 6 - 7 | 986 | 79 | |
BX648848.1 | - | 6 - 7 | 941 | 79 | |
BX648849.1 | - | 6 - 7 | 931 | 79 | |
BX649140.1 | - | 6 - 7 | 987 | 79 | |
CR457053.1 | - | 5 - 6 | 786 | 79 | |
X12451.1 | - | 6 - 7 | 1074 | 79 | |
Y18462.1 | - | 3 - 4 | 411 | 79 |