Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134398.1 | NP_001127870.1 | 27 - 28 | 2394 | 93 |
XM_005272213.1 | XP_005272270.1 | 25 - 26 | 2414 | 93 | |
XM_017015108.1 | XP_016870597.1 | 28 - 29 | 2498 | 93 | |
XM_017015109.1 | XP_016870598.1 | 27 - 28 | 2483 | 93 | |
XM_017015110.1 | XP_016870599.1 | 26 - 27 | 2429 | 93 | |
GenBank mRNA | S76992.1 | - | 27 - 28 | 2353 | 93 |