Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083953.1 | NP_001077422.1 | 23 - 24 | 3853 | 69 |
NM_022065.4 | NP_071348.3 | 23 - 24 | 3641 | 69 | |
NR_073394.1 | 23 - 24 | 3633 | 69 | ||
XM_006712061.3 | XP_006712124.1 | 24 - 25 | 3688 | 69 | |
XM_006712062.1 | XP_006712125.1 | 23 - 24 | 3638 | 69 | |
XM_006712063.1 | XP_006712126.1 | 23 - 24 | 3521 | 69 | |
XM_006712064.2 | XP_006712127.1 | 23 - 24 | 3641 | 69 | |
XM_006712065.1 | XP_006712128.1 | 23 - 24 | 3641 | 69 | |
XM_006712066.1 | XP_006712129.1 | 21 - 22 | 3278 | 69 | |
XM_006712067.1 | XP_006712130.1 | 21 - 22 | 3275 | 69 | |
XM_006712068.3 | XP_006712131.1 | 23 - 24 | 3641 | 69 | |
XM_006712069.3 | XP_006712132.1 | 8 - 9 | 1149 | 69 | |
XM_011533015.2 | XP_011531317.1 | 23 - 24 | 3641 | 69 | |
XM_017004674.1 | XP_016860163.1 | 23 - 24 | 3518 | 69 | |
XM_017004675.1 | XP_016860164.1 | 22 - 23 | 3393 | 69 | |
GenBank mRNA | AB051554.2 | - | 23 - 24 | 3853 | 69 |
AK025445.1 | - | 10 - 11 | 1442 | 69 | |
AK126824.1 | - | 12 - 13 | 2039 | 69 | |
AK290023.1 | - | 23 - 24 | 3641 | 69 | |
AK307915.1 | - | 8 - 9 | 966 | 69 | |
AL832141.1 | - | 23 - 24 | 3623 | 69 | |
AY149629.1 | - | 23 - 24 | 3633 | 69 | |
AY149633.1 | - | 23 - 24 | 3625 | 69 | |
BC105976.1 | - | 2 - 3 | 151 | 69 | |
BX648155.1 | - | 23 - 24 | 3637 | 69 |