Assay Design



Gene Transcripts

Gene Symbol
THADA
Entrez Gene ID
63892
Gene Name
THADA, armadillo repeat containing
Gene Aliases
ARMC13, GITA
Chromosome Location
Chr.2: 43230836 - 43596046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083953.1 NP_001077422.1 21 - 22 3611 92
NM_022065.4 NP_071348.3 21 - 22 3399 92
NR_073394.1 21 - 22 3391 92
XM_006712061.3 XP_006712124.1 22 - 23 3446 92
XM_006712062.1 XP_006712125.1 21 - 22 3396 92
XM_006712063.1 XP_006712126.1 21 - 22 3279 92
XM_006712064.2 XP_006712127.1 21 - 22 3399 92
XM_006712065.1 XP_006712128.1 21 - 22 3399 92
XM_006712066.1 XP_006712129.1 19 - 20 3036 92
XM_006712067.1 XP_006712130.1 19 - 20 3033 92
XM_006712068.3 XP_006712131.1 21 - 22 3399 92
XM_006712069.3 XP_006712132.1 6 - 7 907 92
XM_011533015.2 XP_011531317.1 21 - 22 3399 92
XM_017004674.1 XP_016860163.1 21 - 22 3276 92
XM_017004675.1 XP_016860164.1 20 - 21 3151 92
GenBank mRNA AB051554.2 - 21 - 22 3611 92
AK025445.1 - 8 - 9 1200 92
AK126824.1 - 10 - 11 1797 92
AK290023.1 - 21 - 22 3399 92
AK307915.1 - 6 - 7 724 92
AL832141.1 - 21 - 22 3381 92
AY149629.1 - 21 - 22 3391 92
AY149633.1 - 21 - 22 3383 92
BX648155.1 - 21 - 22 3395 92

View Full Product Details