Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289823.1 | NP_001276752.1 | 13 - 14 | 1786 | 94 |
NM_001289824.1 | NP_001276753.1 | 13 - 14 | 1940 | 94 | |
NM_002569.3 | NP_002560.1 | 13 - 14 | 1839 | 94 | |
GenBank mRNA | BC012181.1 | - | 12 - 13 | 1609 | 94 |
X17094.1 | - | 13 - 14 | 1776 | 94 |