Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 9 - 10 | 875 | 89 |
XM_011534325.2 | XP_011532627.1 | 9 - 10 | 856 | 89 | |
XM_011534327.1 | XP_011532629.1 | 9 - 10 | 856 | 89 | |
XM_011534328.2 | XP_011532630.1 | 9 - 10 | 856 | 89 | |
XM_011534329.1 | XP_011532631.1 | 9 - 10 | 856 | 89 | |
XM_011534330.2 | XP_011532632.1 | 9 - 10 | 856 | 89 | |
XM_017007595.1 | XP_016863084.1 | 7 - 8 | 624 | 89 | |
XM_017007596.1 | XP_016863085.1 | 9 - 10 | 901 | 89 | |
XM_017007598.1 | XP_016863087.1 | 9 - 10 | 856 | 89 | |
XM_017007599.1 | XP_016863088.1 | 9 - 10 | 856 | 89 | |
XM_017007600.1 | XP_016863089.1 | 7 - 8 | 710 | 89 | |
GenBank mRNA | AB015046.1 | - | 9 - 10 | 895 | 89 |
AK001205.1 | - | 2 - 3 | 146 | 89 | |
AK025728.1 | - | 9 - 10 | 827 | 89 | |
AK293325.1 | - | 7 - 8 | 629 | 89 | |
AK314386.1 | - | 9 - 10 | 823 | 89 | |
BC137076.1 | - | 9 - 10 | 807 | 89 | |
BC137080.1 | - | 9 - 10 | 807 | 89 |