Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002931.3 | NP_002922.2 | 3 - 4 | 435 | 69 |
GenBank mRNA | AK289355.1 | - | 3 - 4 | 444 | 69 |
AK301010.1 | - | 3 - 4 | 444 | 69 | |
BC002922.2 | - | 3 - 4 | 430 | 69 | |
BC051866.2 | - | 3 - 4 | 428 | 69 | |
BI259576.1 | - | 3 - 4 | 444 | 69 | |
BT007352.1 | - | 1 - 2 | 154 | 69 | |
Z14000.1 | - | 1 - 2 | 229 | 69 |