Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006411.3 | NP_006402.1 | 4 - 5 | 846 | 103 |
NM_032741.4 | NP_116130.2 | 4 - 5 | 631 | 103 | |
XM_005248805.3 | XP_005248862.1 | 4 - 5 | 1149 | 103 | |
XM_005248806.2 | XP_005248863.1 | 4 - 5 | 1042 | 103 | |
XM_011514234.1 | XP_011512536.1 | 4 - 5 | 813 | 103 | |
GenBank mRNA | AF007145.1 | - | 3 - 4 | 395 | 103 |
AF218034.1 | - | 4 - 5 | 609 | 103 | |
AK058025.1 | - | 5 - 6 | 751 | 103 | |
AK129818.1 | - | 4 - 5 | 592 | 103 | |
AK299252.1 | - | 4 - 5 | 643 | 103 | |
AK315950.1 | - | 3 - 4 | 632 | 103 | |
BC002402.2 | - | 4 - 5 | 585 | 103 | |
BC003007.1 | - | 4 - 5 | 585 | 103 | |
BC004310.1 | - | 4 - 5 | 790 | 103 | |
BC090849.1 | - | 4 - 5 | 802 | 103 | |
U56417.1 | - | 4 - 5 | 829 | 103 | |
U75971.1 | - | 3 - 4 | 520 | 103 | |
Y09565.1 | - | 4 - 5 | 620 | 103 |