Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006411.3 | NP_006402.1 | 6 - 7 | 1018 | 110 |
NM_032741.4 | NP_116130.2 | 6 - 7 | 803 | 110 | |
XM_005248805.3 | XP_005248862.1 | 6 - 7 | 1321 | 110 | |
XM_005248806.2 | XP_005248863.1 | 6 - 7 | 1214 | 110 | |
XM_011514234.1 | XP_011512536.1 | 6 - 7 | 985 | 110 | |
GenBank mRNA | AF007145.1 | - | 5 - 6 | 567 | 110 |
AF218034.1 | - | 6 - 7 | 781 | 110 | |
AK056346.1 | - | 4 - 5 | 1215 | 110 | |
AK058025.1 | - | 7 - 8 | 923 | 110 | |
AK129818.1 | - | 6 - 7 | 764 | 110 | |
AK299252.1 | - | 6 - 7 | 815 | 110 | |
AK315950.1 | - | 5 - 6 | 804 | 110 | |
BC002402.2 | - | 6 - 7 | 757 | 110 | |
BC003007.1 | - | 6 - 7 | 757 | 110 | |
BC004310.1 | - | 6 - 7 | 962 | 110 | |
BC090849.1 | - | 6 - 7 | 974 | 110 | |
U56417.1 | - | 6 - 7 | 1001 | 110 | |
U75971.1 | - | 5 - 6 | 692 | 110 | |
Y09565.1 | - | 6 - 7 | 792 | 110 |