Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321154.1 | NP_001308083.1 | 5 - 6 | 893 | 124 |
NM_001321155.1 | NP_001308084.1 | 6 - 7 | 1017 | 124 | |
NM_001321156.1 | NP_001308085.1 | 6 - 7 | 1087 | 124 | |
NM_001321157.1 | NP_001308086.1 | 6 - 7 | 920 | 124 | |
NM_001321158.1 | NP_001308087.1 | 4 - 5 | 641 | 124 | |
NM_001321159.1 | NP_001308088.1 | 4 - 5 | 738 | 124 | |
NM_001321160.1 | NP_001308089.1 | 6 - 7 | 1017 | 124 | |
NM_001321161.1 | NP_001308090.1 | 4 - 5 | 641 | 124 | |
NM_001321162.1 | NP_001308091.1 | 5 - 6 | 862 | 124 | |
NM_024770.4 | NP_079046.2 | 5 - 6 | 796 | 124 | |
GenBank mRNA | AK024046.1 | - | 5 - 6 | 863 | 124 |
AK124092.1 | - | 6 - 7 | 968 | 124 | |
AK297141.1 | - | 4 - 5 | 124 | ||
BC025250.1 | - | 6 - 7 | 124 |