Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012343.3 | NP_036475.3 | 11 - 12 | 1862 | 65 |
NM_182977.2 | NP_892022.2 | 11 - 12 | 1828 | 65 | |
XM_005248274.4 | XP_005248331.1 | 11 - 12 | 1840 | 65 | |
XM_005248275.4 | XP_005248332.1 | 6 - 7 | 1090 | 65 | |
XM_006714461.3 | XP_006714524.1 | 10 - 11 | 1454 | 65 | |
XM_011514001.2 | XP_011512303.1 | 11 - 12 | 1859 | 65 | |
XM_011514002.2 | XP_011512304.1 | 10 - 11 | 1549 | 65 | |
XM_017009293.1 | XP_016864782.1 | 11 - 12 | 1730 | 65 | |
GenBank mRNA | AK297558.1 | - | 4 - 5 | 475 | 65 |
AL831822.2 | - | 11 - 12 | 1829 | 65 | |
BC110543.1 | - | 11 - 12 | 1678 | 65 | |
BC110544.1 | - | 10 - 11 | 1675 | 65 | |
DA521839.1 | - | 4 - 5 | 475 | 65 | |
U40490.1 | - | 11 - 12 | 1750 | 65 | |
Z50101.1 | - | 10 - 11 | 1650 | 65 |