Assay Design



Gene Transcripts

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
Chromosome Location
Chr.15: 25672241 - 25865144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024490.3 NP_077816.1 14 - 15 3192 62
XM_005268261.4 XP_005268318.1 15 - 16 3322 62
XM_011521826.2 XP_011520128.1 15 - 16 3493 62
XM_011521828.2 XP_011520130.1 14 - 15 3921 62
XM_011521829.2 XP_011520131.1 11 - 12 2325 62
XM_011521830.2 XP_011520132.2 9 - 10 1995 62
XM_017022436.1 XP_016877925.1 14 - 15 2828 62
XM_017022437.1 XP_016877926.1 14 - 15 3192 62
GenBank mRNA AB011138.1 - 9 - 10 2080 62
AB051358.1 - 15 - 16 3285 62
BC052251.1 - 14 - 15 3198 62

View Full Product Details