Assay Design



Gene Transcripts

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
Chromosome Location
Chr.15: 25672241 - 25865144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011521826.2 XP_011520128.1 1 - 2 303 81
GenBank mRNA AB051358.1 - 1 - 2 95 81

View Full Product Details