Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005560.4 | NP_005551.3 | 42 - 43 | 5704 | 85 |
XM_006723796.3 | XP_006723859.1 | 42 - 43 | 5707 | 85 | |
XM_006723798.3 | XP_006723861.1 | 42 - 43 | 5710 | 85 | |
XM_011528818.2 | XP_011527120.1 | 41 - 42 | 5569 | 85 | |
XM_011528819.2 | XP_011527121.1 | 40 - 41 | 5434 | 85 | |
GenBank mRNA | AB067494.1 | - | 39 - 40 | 5048 | 85 |
AF443072.1 | - | 42 - 43 | 5704 | 85 |