Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014616.2 | NP_055431.1 | 3 - 4 | 489 | 90 |
XM_005247241.2 | XP_005247298.1 | 3 - 4 | 488 | 90 | |
XM_005247243.2 | XP_005247300.1 | 1 - 2 | 80 | 90 | |
XM_011512593.2 | XP_011510895.1 | 3 - 4 | 488 | 90 | |
XM_011512594.2 | XP_011510896.1 | 3 - 4 | 488 | 90 | |
XM_017006008.1 | XP_016861497.1 | 1 - 2 | 57 | 90 | |
XM_017006009.1 | XP_016861498.1 | 2 - 3 | 183 | 90 | |
GenBank mRNA | AF156548.1 | - | 3 - 4 | 398 | 90 |
AK296750.1 | - | 3 - 4 | 488 | 90 | |
BC042180.1 | - | 3 - 4 | 489 | 90 |