Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_045204.1 | 2 - 3 | 966 | 69 | |
XM_011542560.2 | XP_011540862.1 | 2 - 3 | 1192 | 69 | |
XM_011542561.2 | XP_011540863.1 | 2 - 3 | 1192 | 69 | |
XM_011542562.2 | XP_011540864.1 | 2 - 3 | 442 | 69 | |
XM_011542563.2 | XP_011540865.1 | 2 - 3 | 507 | 69 | |
GenBank mRNA | AK299783.1 | - | 2 - 3 | 127 | 69 |
AK300082.1 | - | 2 - 3 | 376 | 69 | |
BG468734.1 | - | 2 - 3 | 392 | 69 | |
JF289264.1 | - | 2 - 3 | 168 | 69 |