Assay Design



Gene Transcripts

Gene Symbol
HTR3E
Entrez Gene ID
285242
Gene Name
5-hydroxytryptamine receptor 3E
Gene Aliases
5-HT3-E, 5-HT3E, 5-HT3c1
Chromosome Location
Chr.3: 184097064 - 184106995 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256613.1 NP_001243542.1 6 - 7 1184 83
NM_001256614.1 NP_001243543.1 4 - 5 990 83
NM_182589.2 NP_872395.2 5 - 6 957 83
NM_198313.2 NP_938055.1 5 - 6 1139 83
NM_198314.2 NP_938056.1 4 - 5 912 83
GenBank mRNA AY159813.2 - 5 - 6 957 83
AY349352.1 - 5 - 6 673 83
AY349353.1 - 4 - 5 718 83
BC101182.1 - 5 - 6 904 83
BC101183.2 - 5 - 6 904 83
DQ644022.1 - 6 - 7 1184 83
EU165354.1 - 4 - 5 796 83

View Full Product Details