Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 33 - 34 | 4945 | 63 |
XM_005268629.3 | XP_005268686.1 | 33 - 34 | 5061 | 63 | |
XM_011537877.2 | XP_011536179.1 | 33 - 34 | 5061 | 63 | |
XM_017018787.1 | XP_016874276.1 | 12 - 13 | 1889 | 63 | |
XM_017018788.1 | XP_016874277.1 | 8 - 9 | 1133 | 63 | |
GenBank mRNA | AK131537.1 | - | 8 - 9 | 1249 | 63 |
AL832453.2 | - | 12 - 13 | 2120 | 63 | |
AY792511.1 | - | 33 - 34 | 4824 | 63 | |
BC117180.1 | - | 33 - 34 | 4882 | 63 |