Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 34 - 35 | 5128 | 111 |
XM_005268629.3 | XP_005268686.1 | 34 - 35 | 5244 | 111 | |
XM_011537877.2 | XP_011536179.1 | 34 - 35 | 5244 | 111 | |
XM_017018787.1 | XP_016874276.1 | 13 - 14 | 2072 | 111 | |
XM_017018788.1 | XP_016874277.1 | 9 - 10 | 1316 | 111 | |
GenBank mRNA | AK131537.1 | - | 9 - 10 | 1432 | 111 |
AL832453.2 | - | 13 - 14 | 2303 | 111 | |
AY792511.1 | - | 34 - 35 | 5007 | 111 | |
BC117180.1 | - | 34 - 35 | 5065 | 111 |