Assay Design



Gene Transcripts

Gene Symbol
LRRK2
Entrez Gene ID
120892
Gene Name
leucine rich repeat kinase 2
Gene Aliases
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
Chromosome Location
Chr.12: 40224895 - 40369285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198578.3 NP_940980.3 36 - 37 5434 134
XM_005268629.3 XP_005268686.1 36 - 37 5550 134
XM_011537877.2 XP_011536179.1 36 - 37 5550 134
XM_017018787.1 XP_016874276.1 15 - 16 2378 134
XM_017018788.1 XP_016874277.1 11 - 12 1622 134
GenBank mRNA AK122640.1 - 3 - 4 342 134
AK131537.1 - 11 - 12 1738 134
AL832453.2 - 15 - 16 2609 134
AY792511.1 - 36 - 37 5313 134
BC117180.1 - 36 - 37 5371 134

View Full Product Details