Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 43 - 44 | 6505 | 128 |
XM_005268629.3 | XP_005268686.1 | 43 - 44 | 6621 | 128 | |
XM_011537877.2 | XP_011536179.1 | 43 - 44 | 6621 | 128 | |
XM_017018787.1 | XP_016874276.1 | 22 - 23 | 3449 | 128 | |
XM_017018788.1 | XP_016874277.1 | 18 - 19 | 2693 | 128 | |
GenBank mRNA | AK122640.1 | - | 10 - 11 | 1413 | 128 |
AK131537.1 | - | 18 - 19 | 2809 | 128 | |
AL832453.2 | - | 22 - 23 | 3678 | 128 | |
AY792511.1 | - | 43 - 44 | 6384 | 128 | |
BC117180.1 | - | 43 - 44 | 6442 | 128 |