Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 46 - 47 | 6965 | 149 |
XM_005268629.3 | XP_005268686.1 | 46 - 47 | 7081 | 149 | |
XM_011537877.2 | XP_011536179.1 | 46 - 47 | 7081 | 149 | |
XM_017018787.1 | XP_016874276.1 | 25 - 26 | 3909 | 149 | |
XM_017018788.1 | XP_016874277.1 | 21 - 22 | 3153 | 149 | |
GenBank mRNA | AK122640.1 | - | 13 - 14 | 1873 | 149 |
AK131537.1 | - | 21 - 22 | 3269 | 149 | |
AL832453.2 | - | 25 - 26 | 4138 | 149 | |
AL834529.1 | - | 3 - 4 | 463 | 149 | |
AY792511.1 | - | 46 - 47 | 6844 | 149 | |
BC117180.1 | - | 46 - 47 | 6902 | 149 |