Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301065.1 | NP_001287994.1 | 10 - 11 | 1266 | 92 |
NM_007180.2 | NP_009111.2 | 11 - 12 | 1359 | 92 | |
GenBank mRNA | AB000824.1 | - | 11 - 12 | 1369 | 92 |
AK091981.1 | - | 9 - 10 | 1228 | 92 | |
AK223140.1 | - | 11 - 12 | 1333 | 92 | |
AK223143.1 | - | 11 - 12 | 1337 | 92 | |
BC109206.2 | - | 10 - 11 | 1260 | 92 |