Assay Design



Gene Transcripts

Gene Symbol
TREH
Entrez Gene ID
11181
Gene Name
trehalase
Gene Aliases
TRE, TREA
Chromosome Location
Chr.11: 118658232 - 118679672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007180.2 NP_009111.2 5 - 6 570 84
GenBank mRNA AB000824.1 - 5 - 6 580 84
AK223140.1 - 5 - 6 544 84
AK223143.1 - 5 - 6 548 84
AK309565.1 - 5 - 6 544 84

View Full Product Details