Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003642.3 | NP_003633.1 | 4 - 5 | 380 | 125 |
NR_027862.1 | 3 - 4 | 304 | 125 | ||
XM_006712808.3 | XP_006712871.1 | 5 - 6 | 576 | 125 | |
XM_011512027.2 | XP_011510329.1 | 4 - 5 | 1132 | 125 | |
XM_017005136.1 | XP_016860625.1 | 5 - 6 | 1353 | 125 | |
XM_017005137.1 | XP_016860626.1 | 5 - 6 | 1285 | 125 | |
GenBank mRNA | AF030424.1 | - | 4 - 5 | 340 | 125 |
AK313699.1 | - | 4 - 5 | 371 | 125 | |
BC063003.1 | - | 3 - 4 | 281 | 125 |