Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289395.1 | NP_001276324.1 | 5 - 6 | 486 | 95 |
NM_003322.4 | NP_003313.3 | 6 - 7 | 645 | 95 | |
GenBank mRNA | AB593123.1 | - | 6 - 7 | 645 | 95 |
AB593124.1 | - | 6 - 7 | 631 | 95 | |
BC032714.1 | - | 5 - 6 | 471 | 95 | |
BC065261.1 | - | 5 - 6 | 454 | 95 | |
U82468.1 | - | 6 - 7 | 637 | 95 |