Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005184.2 | NP_005175.2 | 4 - 5 | 460 | 79 |
GenBank mRNA | AK295927.1 | - | 4 - 5 | 337 | 79 |
AK310762.1 | - | 5 - 6 | 362 | 79 | |
BC005137.2 | - | 4 - 5 | 364 | 79 | |
BC006182.1 | - | 4 - 5 | 565 | 79 | |
BG753029.1 | - | 4 - 5 | 461 | 79 | |
BT006855.1 | - | 3 - 4 | 280 | 79 | |
FJ224310.1 | - | 4 - 5 | 383 | 79 | |
J04046.1 | - | 4 - 5 | 383 | 79 |