Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144891.1 | NP_001138363.1 | 3 - 4 | 568 | 113 |
NM_001308110.1 | NP_001295039.1 | 3 - 4 | 568 | 113 | |
NM_016586.2 | NP_057670.2 | 3 - 4 | 568 | 113 | |
XM_005267754.3 | XP_005267811.1 | 3 - 4 | 563 | 113 | |
XM_005267756.4 | XP_005267813.1 | 3 - 4 | 563 | 113 | |
XM_017021367.1 | XP_016876856.1 | 3 - 4 | 563 | 113 | |
GenBank mRNA | AB038523.1 | - | 3 - 4 | 537 | 113 |
AF208857.1 | - | 3 - 4 | 517 | 113 | |
AK315290.1 | - | 3 - 4 | 569 | 113 | |
BC005197.1 | - | 3 - 4 | 527 | 113 | |
BC016821.1 | - | 3 - 4 | 523 | 113 | |
BX248287.1 | - | 3 - 4 | 488 | 113 |