Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016586.2 | NP_057670.2 | 7 - 8 | 974 | 82 |
XM_005267754.3 | XP_005267811.1 | 6 - 7 | 903 | 82 | |
GenBank mRNA | AB038523.1 | - | 7 - 8 | 943 | 82 |
AK293473.1 | - | 7 - 8 | 855 | 82 | |
AK315290.1 | - | 7 - 8 | 975 | 82 |