Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270521.1 | NP_001257450.1 | 16 - 17 | 2119 | 86 |
NM_001270522.1 | NP_001257451.1 | 16 - 17 | 2119 | 86 | |
NM_004830.3 | NP_004821.2 | 16 - 17 | 2119 | 86 | |
NM_015979.3 | NP_057063.2 | 17 - 18 | 2137 | 86 | |
XM_005267223.2 | XP_005267280.1 | 17 - 18 | 2133 | 86 | |
XM_006715612.2 | XP_006715675.1 | 17 - 18 | 2133 | 86 | |
XM_011536257.2 | XP_011534559.1 | 15 - 16 | 2019 | 86 | |
XM_017011501.1 | XP_016866990.1 | 7 - 8 | 1077 | 86 | |
GenBank mRNA | AB033042.1 | - | 16 - 17 | 2088 | 86 |
AF104255.1 | - | 17 - 18 | 2097 | 86 | |
AF105332.1 | - | 16 - 17 | 2048 | 86 | |
AF135022.1 | - | 16 - 17 | 1929 | 86 | |
AK304711.1 | - | 5 - 6 | 858 | 86 | |
AK315030.1 | - | 16 - 17 | 2000 | 86 | |
AL136776.1 | - | 17 - 18 | 2103 | 86 | |
BC060759.1 | - | 16 - 17 | 2063 | 86 | |
EU392520.1 | - | 8 - 9 | 818 | 86 | |
EU392521.1 | - | 1 - 2 | 57 | 86 | |
EU392522.1 | - | 4 - 5 | 358 | 86 | |
EU392523.1 | - | 1 - 2 | 57 | 86 | |
EU392524.1 | - | 1 - 2 | 57 | 86 | |
EU392526.1 | - | 14 - 15 | 1551 | 86 | |
EU392527.1 | - | 7 - 8 | 837 | 86 | |
EU392528.1 | - | 6 - 7 | 556 | 86 | |
EU392529.1 | - | 6 - 7 | 447 | 86 | |
EU392530.1 | - | 5 - 6 | 426 | 86 | |
EU392531.1 | - | 6 - 7 | 417 | 86 | |
EU392532.1 | - | 3 - 4 | 435 | 86 |