Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270521.1 | NP_001257450.1 | 8 - 9 | 858 | 84 |
NM_001270522.1 | NP_001257451.1 | 8 - 9 | 858 | 84 | |
NM_004830.3 | NP_004821.2 | 8 - 9 | 858 | 84 | |
NM_015979.3 | NP_057063.2 | 8 - 9 | 858 | 84 | |
XM_005267223.2 | XP_005267280.1 | 8 - 9 | 854 | 84 | |
XM_006715612.2 | XP_006715675.1 | 8 - 9 | 854 | 84 | |
XM_011536257.2 | XP_011534559.1 | 8 - 9 | 854 | 84 | |
GenBank mRNA | AB033042.1 | - | 8 - 9 | 827 | 84 |
AF104255.1 | - | 8 - 9 | 818 | 84 | |
AF105332.1 | - | 8 - 9 | 787 | 84 | |
AF135022.1 | - | 8 - 9 | 668 | 84 | |
AK315030.1 | - | 8 - 9 | 739 | 84 | |
AL136776.1 | - | 8 - 9 | 824 | 84 | |
BC060759.1 | - | 8 - 9 | 802 | 84 | |
EU392526.1 | - | 8 - 9 | 679 | 84 |