Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321851.1 | NP_001308780.1 | 10 - 11 | 1901 | 114 |
NM_004239.4 | NP_004230.2 | 10 - 11 | 1904 | 114 | |
XM_017021787.1 | XP_016877276.1 | 11 - 12 | 1925 | 114 | |
XM_017021788.1 | XP_016877277.1 | 8 - 9 | 1776 | 114 | |
GenBank mRNA | AK307489.1 | - | 10 - 11 | 1904 | 114 |
BC146845.1 | - | 10 - 11 | 1786 | 114 | |
BX641024.1 | - | 4 - 5 | 676 | 114 | |
Y12490.1 | - | 10 - 11 | 1886 | 114 |