Assay Design



Gene Transcripts

Gene Symbol
TRIP11
Entrez Gene ID
9321
Gene Name
thyroid hormone receptor interactor 11
Gene Aliases
ACG1A, CEV14, GMAP-210, GMAP210, TRIP-11, TRIP230
Chromosome Location
Chr.14: 91965991 - 92040140 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321851.1 NP_001308780.1 3 - 4 683 83
NM_004239.4 NP_004230.2 3 - 4 686 83
XM_017021787.1 XP_016877276.1 3 - 4 666 83
XM_017021788.1 XP_016877277.1 3 - 4 686 83
GenBank mRNA AK307489.1 - 3 - 4 686 83
BC146845.1 - 3 - 4 568 83
Y12490.1 - 3 - 4 668 83

View Full Product Details