Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321851.1 | NP_001308780.1 | 4 - 5 | 955 | 137 |
NM_004239.4 | NP_004230.2 | 4 - 5 | 958 | 137 | |
XM_017021787.1 | XP_016877276.1 | 4 - 5 | 938 | 178 | |
XM_017021788.1 | XP_016877277.1 | 4 - 5 | 958 | 137 | |
GenBank mRNA | AF007217.1 | - | 4 - 5 | 935 | 137 |
AK307489.1 | - | 4 - 5 | 958 | 137 | |
BC146845.1 | - | 4 - 5 | 840 | 137 | |
Y12490.1 | - | 4 - 5 | 940 | 137 |