Assay Design
Gene Transcripts
- Gene Symbol
- PARD3
- Entrez Gene ID
-
56288
- Gene Name
- par-3 family cell polarity regulator
- Gene Aliases
-
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
- Chromosome Location
-
Chr.10: 34109560 - 34815325 on Build GRCh38
View Full Product Details