Assay Design



Gene Transcripts

Gene Symbol
PARD3
Entrez Gene ID
56288
Gene Name
par-3 family cell polarity regulator
Gene Aliases
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location
Chr.10: 34109560 - 34815325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184785.1 NP_001171714.1 16 - 17 2737 75
NM_001184786.1 NP_001171715.1 15 - 16 2698 75
NM_001184787.1 NP_001171716.1 16 - 17 2746 75
NM_001184788.1 NP_001171717.1 15 - 16 2698 75
NM_001184789.1 NP_001171718.1 15 - 16 2698 75
NM_001184790.1 NP_001171719.1 14 - 15 2566 75
NM_001184791.1 NP_001171720.1 14 - 15 2566 75
NM_001184792.1 NP_001171721.1 16 - 17 2737 75
NM_001184793.1 NP_001171722.1 16 - 17 2737 75
NM_001184794.1 NP_001171723.1 15 - 16 2698 75
NM_019619.3 NP_062565.2 16 - 17 2746 75
XM_005252528.4 XP_005252585.1 16 - 17 2728 75
XM_005252530.4 XP_005252587.1 16 - 17 2728 75
XM_005252531.4 XP_005252588.1 16 - 17 2728 75
XM_005252532.4 XP_005252589.1 15 - 16 2382 75
XM_005252534.4 XP_005252591.1 16 - 17 2728 75
XM_005252535.4 XP_005252592.1 15 - 16 2689 75
XM_011519569.2 XP_011517871.1 16 - 17 2737 75
XM_011519570.2 XP_011517872.1 15 - 16 2698 75
XM_011519571.2 XP_011517873.1 16 - 17 2737 75
XM_011519572.2 XP_011517874.1 16 - 17 2737 75
XM_011519573.2 XP_011517875.1 16 - 17 2737 75
XM_011519575.2 XP_011517877.1 15 - 16 2421 75
XM_011519576.2 XP_011517878.1 15 - 16 2392 75
XM_011519577.2 XP_011517879.1 14 - 15 2569 75
XM_011519578.2 XP_011517880.1 14 - 15 2570 75
XM_011519579.2 XP_011517881.1 14 - 15 2290 75
XM_011519581.2 XP_011517883.1 16 - 17 2737 75
XM_011519583.2 XP_011517885.1 16 - 17 2737 75
XM_017016430.1 XP_016871919.1 16 - 17 2728 75
GenBank mRNA AA136967.1 - 2 - 3 304 75
AB073671.1 - 14 - 15 2236 75
AF177228.1 - 9 - 10 1482 75
AF196185.1 - 16 - 17 2694 75
AF196186.1 - 16 - 17 2694 75
AF252293.1 - 14 - 15 2236 75
AF332592.1 - 15 - 16 2368 75
AF332593.1 - 15 - 16 2368 75
AF454057.1 - 14 - 15 2114 75
AF454058.1 - 14 - 15 2589 75
AF454059.1 - 11 - 12 1701 75
AF467002.1 - 16 - 17 2416 75
AF467003.1 - 16 - 17 2407 75
AF467004.1 - 16 - 17 2416 75
AF467005.1 - 14 - 15 2236 75
AF467006.1 - 15 - 16 2368 75
AK024668.1 - 3 - 4 389 75
AK025892.1 - 7 - 8 1184 75
AK027735.1 - 14 - 15 2034 75
BC071566.1 - 15 - 16 2645 75

View Full Product Details