Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130852.1 | NP_001124324.1 | 3 - 4 | 725 | 71 |
NM_012116.3 | NP_036248.3 | 3 - 4 | 725 | 71 | |
XM_005258696.3 | XP_005258753.1 | 3 - 4 | 732 | 71 | |
XM_011526688.2 | XP_011524990.1 | 3 - 4 | 733 | 71 | |
XM_011526689.2 | XP_011524991.1 | 3 - 4 | 735 | 71 | |
GenBank mRNA | AB028645.1 | - | 3 - 4 | 703 | 71 |
AF117646.1 | - | 3 - 4 | 725 | 71 | |
AF117647.1 | - | 3 - 4 | 725 | 71 | |
BC028915.1 | - | 3 - 4 | 683 | 71 |